missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 4-bromotetrahydro-1(2H)-pyridinecarboxylate, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Density | 1.374g/mL |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 298.18 |
Percent Purity | 97% |
Chemical Name or Material | Benzyl 4-bromotetrahydro-1(2H)-pyridinecarboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'