missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-bromophenyl ketone, Thermo Scientific Chemicals

Catalog Number p-3756893
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2001-29-8
C14H11BrO
275.14
MFCD00016331
MOSIKPSTRPODHQ-UHFFFAOYSA-N
benzyl 4-bromophenyl ketone, 1-4-bromophenyl-2-phenylethanone, 4'-bromo-2-phenylacetophenone, 4-bromodeoxybenzoin, 4-bromodesoxybenzoin, 1-4-bromophenyl-2-phenyl-ethanone, ethanone, 1-4-bromophenyl-2-phenyl, 1-4-bromophenyl-2-phenylethan-1-one, acmc-1chtj
519738
1-(4-bromophenyl)-2-phenylethanone
C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br
This item is not returnable. View return policy
2001-29-8
C14H11BrO
275.14
MFCD00016331
MOSIKPSTRPODHQ-UHFFFAOYSA-N
benzyl 4-bromophenyl ketone, 1-4-bromophenyl-2-phenylethanone, 4'-bromo-2-phenylacetophenone, 4-bromodeoxybenzoin, 4-bromodesoxybenzoin, 1-4-bromophenyl-2-phenyl-ethanone, ethanone, 1-4-bromophenyl-2-phenyl, 1-4-bromophenyl-2-phenylethan-1-one, acmc-1chtj
519738
1-(4-bromophenyl)-2-phenylethanone
C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 109°C to 112°C
Color White to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH2COC6H4Br
Quantity 1 g
Formula Weight 275.14
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 4-bromophenyl ketone
compliance-icons
Product Identifier
  • Benzyl 4-bromophenyl ketone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'