missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 3-pyrroline-1-carboxylate, 98%, Thermo Scientific Chemicals

Catalog Number p-4229086
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
31970-04-4
C12H13NO2
203.24
XSKKIFJNZPNVGO-UHFFFAOYSA-N
benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester
643471
benzyl 2,5-dihydropyrrole-1-carboxylate
C1C=CCN1C(=O)OCC2=CC=CC=C2
This item is not returnable. View return policy
31970-04-4
C12H13NO2
203.24
XSKKIFJNZPNVGO-UHFFFAOYSA-N
benzyl 2,5-dihydro-1h-pyrrole-1-carboxylate, benzyl 3-pyrroline-1-carboxylate, 1-cbz-2,5-dihydro-1h-pyrrole, n-cbz-2,5-dihydropyrrole, n-cbz-3-pyrroline, n-carbobenzoxy-3-pyrroline, 2,5-dihydropyrrole-1-carboxylic acid benzyl ester, benzyl 2h-pyrrole-1 5h-carboxylate, n-benzyloxycarbonyl-3-pyrroline, 1h-pyrrole-1-carboxylic acid, 2,5-dihydro-, phenylmethyl ester
643471
benzyl 2,5-dihydropyrrole-1-carboxylate
C1C=CCN1C(=O)OCC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 133.0°C to 137.0°C (0.6mmHg)
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Assay Percent Range 97.5% min. (GC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.544
Formula Weight 203.24
Percent Purity 98%
Density 1.1320g/mL
Quantity 1 g
Flash Point >110°C
Specific Gravity 1.132
Physical Form Liquid
Chemical Name or Material Benzyl 3-pyrroline-1-carboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl 3-pyrroline-1-carboxylate, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'