missing translation for 'onlineSavingsMsg'
Learn More

(Benzoylmethylene)triphenylphosphorane, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081970
Click to view available options
Quantity:
5 g
25 g
100 g
859-65-4
C26H21OP
380.43
MFCD00014088
MZRSAJZDYIISJW-UHFFFAOYSA-N
benzoylmethylene triphenylphosphorane, 2-triphenylphosphoranylidene acetophenone, 1-phenyl-2-triphenylphosphoranylidene ethanone, phenacylidenetriphenyl phosphorane, phenylcarbonylmethylene triphenylphosphorane, acetophenone, 2-triphenylphosphoranylidene, phenacylidenetriphenylphosphorane, triphenylphosphinebenzoylmethylene, ethanone, 1-phenyl-2-triphenylphosphoranylidene
70073
1-phenyl-2-(triphenyl-$l^{5}-phosphanylidene)ethanone
O=C(C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
859-65-4
C26H21OP
380.43
MFCD00014088
MZRSAJZDYIISJW-UHFFFAOYSA-N
benzoylmethylene triphenylphosphorane, 2-triphenylphosphoranylidene acetophenone, 1-phenyl-2-triphenylphosphoranylidene ethanone, phenacylidenetriphenyl phosphorane, phenylcarbonylmethylene triphenylphosphorane, acetophenone, 2-triphenylphosphoranylidene, phenacylidenetriphenylphosphorane, triphenylphosphinebenzoylmethylene, ethanone, 1-phenyl-2-triphenylphosphoranylidene
70073
1-phenyl-2-(triphenyl-$l^{5}-phosphanylidene)ethanone
O=C(C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

It is used in organic synthesis, wittig reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic synthesis, wittig reagent.

Solubility
Soluble in Methanol (almost transparency).

Notes
Air sensitive. Store in refrigerator. Store away from oxidizing agent, air.
TRUSTED_SUSTAINABILITY
Melting Point 183°C to 186°C
Quantity 5 g
Beilstein 620285
Sensitivity Air sensitive
Solubility Information Soluble in Methanol (almost transparency).
Formula Weight 380.43
Percent Purity ≥98%
Chemical Name or Material (Benzoylmethylene)triphenylphosphorane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'