missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl isothiocyanate, 98%, Thermo Scientific Chemicals

Catalog Number p-3756885
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
532-55-8
C8H5NOS
163.2
MFCD00004815
CPEKAXYCDKETEN-UHFFFAOYSA-N
benzoylthiocarbimide, benzoylisothiocyanate, benzoylisothiocyante, isothiocyanic acid benzoyl ester, unii-u8j7q32b86, n-benzoyl isothiocyanate, benzoic acid, anhydride with hncs, benzoic acid, anhydride with isothiocyanic acid, oxophenylmethanisothiocyanate, 1-benzoyl acetone
68284
benzoyl isothiocyanate
C1=CC=C(C=C1)C(=O)N=C=S
This item is not returnable. View return policy
532-55-8
C8H5NOS
163.2
MFCD00004815
CPEKAXYCDKETEN-UHFFFAOYSA-N
benzoylthiocarbimide, benzoylisothiocyanate, benzoylisothiocyante, isothiocyanic acid benzoyl ester, unii-u8j7q32b86, n-benzoyl isothiocyanate, benzoic acid, anhydride with hncs, benzoic acid, anhydride with isothiocyanic acid, oxophenylmethanisothiocyanate, 1-benzoyl acetone
68284
benzoyl isothiocyanate
C1=CC=C(C=C1)C(=O)N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Density 1.2100g/mL
Boiling Point 128.0°C to 131.0°C (15.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CONCS
Refractive Index 1.6350 to 1.6370
Quantity 5 g
Beilstein 09, IV, 777
Merck Index 15, 1117
Specific Gravity 1.21
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in chloroform,carbon tetrachloride,,dioxane
Formula Weight 163.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzoyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'