missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoyl chloride, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.
Solubility
Miscible with water, ether, benzene, carbon disulfide and carbon tetra chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases and alcohols.
Benzoyl chloride is widely utilized for the synthesis of peroxides. It is employed in the production of dyes and perfumes. It also serves in the manufacturing of pharmaceuticals and resins.
Solubility
Miscible with water, ether, benzene, carbon disulfide and carbon tetra chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong bases and alcohols.
Specifications
Melting Point | -1°C |
pH | 2 |
Boiling Point | 197°C to 199°C |
Flash Point | 69°C (156°F) |
Assay Percent Range | ≥99% |
Odor | Pungent |
Quantity | 5000 g |
UN Number | UN1736 |
Merck Index | 14,1112 |
SMILES | C1=CC=C(C=C1)C(=O)Cl |
Show More |
Safety and Handling
Product Identifier
- Benzoyl chloride
- Danger
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'