missing translation for 'onlineSavingsMsg'
Learn More

(Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate, 98%, Thermo Scientific™

Catalog Number 11916401
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
190849-64-0
C17H24F6N5OP
459.38
MFCD00191769
YNOBMGHLCWIWCL-UHFFFAOYSA-N
hbpipu, benzotriazol-1-yloxy dipiperidinocarbenium hexafluorophosphate, 1-1h-benzo d 1,2,3 triazol-1-yl oxy piperidin-1-yl methylene piperidin-1-ium hexafluorophosphate v, hbpipu, benzotriazol-1-yloxy dipiperidinocarbenium hexafluorophosphate, o-benzotriazol-1-yl-n,n,n',n'-bis pentamethylene uronium hexafluorophosphate, 1,2,3-benzotriazol-1-yloxy bis piperidin-1-yl methylium hexafluorophosphate, 1-piperidin-1-ium-1-ylidene 1-piperidyl methoxy benzotriazole hexafluorophosphate, 1-bis 1-piperidyl methoxy benzotriazole hexafluorophosphate, o-benzotriazol-1-yl-n,n,n',n'-bis pentamethylene uroniumhexafluorophosphate, 1,2,3-benzotriazol-1-yl bis piperidin-1-yl methylidene oxidanium hexafluorophosphate
10863473
1-[piperidin-1-ium-1-ylidene(piperidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
F[P-](F)(F)(F)(F)F.C1CCN(CC1)C(ON1N=NC2=CC=CC=C12)=[N+]1CCCCC1
This item is not returnable. View return policy
190849-64-0
C17H24F6N5OP
459.38
MFCD00191769
YNOBMGHLCWIWCL-UHFFFAOYSA-N
hbpipu, benzotriazol-1-yloxy dipiperidinocarbenium hexafluorophosphate, 1-1h-benzo d 1,2,3 triazol-1-yl oxy piperidin-1-yl methylene piperidin-1-ium hexafluorophosphate v, hbpipu, benzotriazol-1-yloxy dipiperidinocarbenium hexafluorophosphate, o-benzotriazol-1-yl-n,n,n',n'-bis pentamethylene uronium hexafluorophosphate, 1,2,3-benzotriazol-1-yloxy bis piperidin-1-yl methylium hexafluorophosphate, 1-piperidin-1-ium-1-ylidene 1-piperidyl methoxy benzotriazole hexafluorophosphate, 1-bis 1-piperidyl methoxy benzotriazole hexafluorophosphate, o-benzotriazol-1-yl-n,n,n',n'-bis pentamethylene uroniumhexafluorophosphate, 1,2,3-benzotriazol-1-yl bis piperidin-1-yl methylidene oxidanium hexafluorophosphate
10863473
1-[piperidin-1-ium-1-ylidene(piperidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
F[P-](F)(F)(F)(F)F.C1CCN(CC1)C(ON1N=NC2=CC=CC=C12)=[N+]1CCCCC1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 183°C to 190°C
Color White
Elemental Analysis 43.34 to 45.56% C, 14.86 to 15.63% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 1 g
Formula Weight 459.38
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • (Benzotriazol-1-yloxy)dipiperidinocarbenium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'