missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7080731
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
95-16-9
C7H5NS
135.184
MFCD00005775
IOJUPLGTWVMSFF-UHFFFAOYSA-N
benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
7222
CHEBI:45993
1,3-benzothiazole
C1=CC=C2C(=C1)N=CS2
This item is not returnable. View return policy
95-16-9
C7H5NS
135.184
MFCD00005775
IOJUPLGTWVMSFF-UHFFFAOYSA-N
benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
7222
CHEBI:45993
1,3-benzothiazole
C1=CC=C2C(=C1)N=CS2

It is an interesting carbonyl equivalent and reacts with aldehydes or ketones to generate α-hydroxy carbonyl compounds. Benzothiazole is used as a chemical intermediate in organic synthesis. It is a precursor of rubber accelerators and a component of cyanine dyes. It is also used as a flavoring substance. It is also used as a flavor, antimicrobial agent, and component of cyanine dyes. Its derivatives are used as rubber accelerators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an interesting carbonyl equivalent and reacts with aldehydes or ketones to generate α-hydroxy carbonyl compounds. Benzothiazole is used as a chemical intermediate in organic synthesis. It is a precursor of rubber accelerators and a component of cyanine dyes. It is also used as a flavoring substance. It has been used as an antimicrobial agent in athletic shoe insoles. It is also used as a flavor, antimicrobial agent, and component of cyanine dyes. Its derivatives are used as rubber accelerators.

Solubility
Slightly soluble in water. Soluble in alcohol, carbon disulfide

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 2°C
Density 1.241
Boiling Point 230°C
Flash Point 107°C (224°F)
Odor Characteristic
Refractive Index 1.642
Quantity 50 g
UN Number UN2810
Beilstein 109468
Merck Index 14,1107
Solubility Information Slightly soluble in water; Soluble in alcohol, carbon disulfide
Formula Weight 135.19
Percent Purity 97%
Chemical Name or Material Benzothiazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'