missing translation for 'onlineSavingsMsg'
Learn More

Benzoquinonium dibromide, Tocris Bioscience™

Nicotinic antagonist

Brand:  Tocris (Bio-Techne) 042410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13467633

  • £89.67 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Nicotinic antagonist, ganglion blocker. Activates single channels but blocks open channels at nicotinic receptors.
Specifications

Chemical Identifiers

311-09-1
706.608
benzoquinonium dibromide, n,n'-3,6-dioxo-1,4-cyclohexadiene-1,4-diyl bis imino-3,1-propanediyl bis n,n-diethylbenzenemethanaminium dibromide
benzyl-[3-[[4-[3-[benzyl(diethyl)azaniumyl]propylamino]-3,6-dioxocyclohexa-1,4-dien-1-yl]amino]propyl]-diethylazanium;dibromide
C34H50Br2N4O2
DVQZHWAYOLXTHE-UHFFFAOYSA-N
56972152
CC[N+](CC)(CCCNC1=CC(=O)C(=CC1=O)NCCC[N+](CC)(CC)CC2=CC=CC=C2)CC3=CC=CC=C3.[Br-].[Br-]

Specifications

10mg
311-09-1
benzoquinonium dibromide, n,n'-3,6-dioxo-1,4-cyclohexadiene-1,4-diyl bis imino-3,1-propanediyl bis n,n-diethylbenzenemethanaminium dibromide
CC[N+](CC)(CCCNC1=CC(=O)C(=CC1=O)NCCC[N+](CC)(CC)CC2=CC=CC=C2)CC3=CC=CC=C3.[Br-].[Br-]
706.608
706.6
Benzoquinonium dibromide
C34H50Br2N4O2
DVQZHWAYOLXTHE-UHFFFAOYSA-N
benzyl-[3-[[4-[3-[benzyl(diethyl)azaniumyl]propylamino]-3,6-dioxocyclohexa-1,4-dien-1-yl]amino]propyl]-diethylazanium;dibromide
56972152
>98%
Videos
Documents