missing translation for 'onlineSavingsMsg'
Learn More

Benzopinacol, 98%, Thermo Scientific Chemicals

Catalog Number p-7122664
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
464-72-2
C26H22O2
366.46
MFCD00004448
MFEWNFVBWPABCX-UHFFFAOYSA-N
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
94766
1,1,2,2-tetraphenylethane-1,2-diol
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O
This item is not returnable. View return policy
464-72-2
C26H22O2
366.46
MFCD00004448
MFEWNFVBWPABCX-UHFFFAOYSA-N
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
94766
1,1,2,2-tetraphenylethane-1,2-diol
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O

Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications:  Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.
TRUSTED_SUSTAINABILITY
Melting Point 174°C to 180°C
Quantity 25 g
Beilstein 2061947
Merck Index 14,1100
Solubility Information It is soluble in ethanol,ether,acetone insoluble in carbon disulfide and water.
Formula Weight 366.46
Percent Purity 98%
Chemical Name or Material Benzopinacol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'