missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone, 99%, pure, Thermo Scientific Chemicals

Catalog Number p-4229081
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
119-61-9
C13H10O
182.22
RWCCWEUUXYIKHB-UHFFFAOYSA-N
benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
3102
CHEBI:41308
diphenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2
119-61-9
C13H10O
182.22
RWCCWEUUXYIKHB-UHFFFAOYSA-N
benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
3102
CHEBI:41308
diphenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 47.0°C to 49.0°C
Color White
Boiling Point 305.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5)2CO
Quantity 500 g
Beilstein 07,411
Fieser 13,108
Merck Index 15,11
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/7.5mL alcohol,1g/6mL ether,soluble in chloroform,acetone and benzene
Formula Weight 182.22
Percent Purity 99%
Grade Pure
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Benzophenone
Show More Show Less
compliance-icons
Product Identifier
  • Benzophenone
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'