missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoic Acid, Certified AR for Analysis, Fisher Chemical™
35.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
35.96 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
35.96 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
1 kg
Description
Benzoic Acid, Certified AR for Analysis
Specifications
Melting Point | 121°C |
Color | White |
pH | 2.5 to 3.5 |
Boiling Point | 249°C |
Packaging | Plastic powder jar |
Quantity | 100 g |
Formula Weight | 122.12g/mol |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Benzoic acid
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 1
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'