missing translation for 'onlineSavingsMsg'
Learn More

Benzoic Acid, Certified AR for Analysis, Fisher Chemical™

Catalog Number 10172180
Click to view available options
Quantity:
100 g
500 g
1 kg
65-85-0
C7H6O2
2398
65-85-0
C7H6O2
2398

Benzoic Acid, Certified AR for Analysis

TRUSTED_SUSTAINABILITY
Melting Point 121°C
Color White
pH 2.5 to 3.5
Boiling Point 249°C
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 122.12g/mol
Physical Form Solid
compliance-icons compliance-icons
Product Identifier
  • Benzoic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'