missing translation for 'onlineSavingsMsg'
Learn More

Benzoic acid, 99%, extra pure, Thermo Scientific Chemicals

Catalog Number p-3586397
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
This item is not returnable. View return policy
65-85-0
C7H6O2
122.12
MFCD00002398
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
243
CHEBI:30746
benzoic acid
C1=CC=C(C=C1)C(=O)O
This item is not returnable. View return policy
65-85-0
C7H6O2
122.12
MFCD00002398
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
243
CHEBI:30746
benzoic acid
C1=CC=C(C=C1)C(=O)O

Benzoic acid, 99%, C7H6O2, CAS Number-65-85-0, benzenecarboxylic acid, benzeneformic acid, benzenemethanoic acid, benzoesaeure gk, benzoesaeure gv, carboxybenzene, dracylic acid, phenylcarboxylic acid, phenylformic acid, retardex, 2.5kg, (20 wt.% soln. in ethanol) clear, 09,92, 249.0 deg.C, 100.0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 121.0°C to 123.0°C
Color Orange to White
Boiling Point 249.0°C
Flash Point 121°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CO2H
Quantity 500 g
Beilstein 09,92
Fieser 01,49; 13,312
Merck Index 15,1093
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in alcohol,benzene,ether,chloroform,,ccl4,carbon disulfide and turpentine
Formula Weight 122.12
Clarity (20 wt.% soln. in ethanol) clear
Percent Purity 99%
Grade Extra Pure
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Benzoic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzoic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'