Learn More
Benzohydroxamic acid, 98%, Thermo Scientific Chemicals
Description
Benzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.
Solubility
Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∼125°C (decomposition) |
Quantity | 10 g |
Beilstein | 1907585 |
Solubility Information | Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene. |
Formula Weight | 137.14 |
Percent Purity | 98% |
Chemical Name or Material | Benzohydroxamic acid |
RUO – Research Use Only