missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Benzohydroxamic acid, 98%

Brand:  Alfa Aesar™ A10509.30

Code : NEW

Additional Details : CAS Number : 495-18-1 Weight : 0.25000kg

Quantity 250g

 View more versions of this product

Product Code. 11448763

  • £895.28 / 250g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Description

Description

Applications
Benzhydroxamic acid is used as precursor in the synthesis of novel mono-anionic and di-anionic hydroxamato complexes by reacting with BiPh3 and Bi(O(t)Bu)3, which has anti-bacterial activity against helicobacter pylori. It is used in the photometric determination of trace amounts of vanadium in alloy steels by making mixed-ligand vanadium chelates with ammonium thiocyanate. It is also involved in the palladium catalyzed synthesis of benzisoxazolones.

Solubility
Soluble in alcohol. Slightly soluble in ether. Insoluble in benzene.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

Benzohydroxamic acid
C7H7NO2
MFCD00002109
Benzhydroxamic acid; N-Benzoylhydroxylamine
495-18-1
137.14
1907585

Specifications

Benzohydroxamic acid
98%
137.14
1907585
≈125°C (dec.)
495-18-1
C7H7NO2
MFCD00002109
Benzhydroxamic acid; N-Benzoylhydroxylamine
250g
Videos
SDS