missing translation for 'onlineSavingsMsg'
Learn More

Benzo[d]isoxazol-3-ol, ≥95%, Thermo Scientific™

Catalog Number p-2703689
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
21725-69-9
C7H5NO2
135.122
MFCD00125030
QLDQYRDCPNBPII-UHFFFAOYSA-N
benzo d isoxazol-3-ol, 1,2-benzisoxazol-3 2h-one, 1,2-benzoxazol-3-ol, 1,2-benzisoxazol-3-ol, 3-hydroxybenzisoxazole, 3-hydroxy-1,2-benzisoxazole, 1,2-benzisoxazolin-3-one, 1,2-benzisoxazol-3-ol 8ci, 2h-1,2-benzoxazol-3-one, benzisoxazolone
210830
1,2-benzoxazol-3-one
C1=CC=C2C(=C1)C(=O)NO2
This item is not returnable. View return policy
21725-69-9
C7H5NO2
135.122
MFCD00125030
QLDQYRDCPNBPII-UHFFFAOYSA-N
benzo d isoxazol-3-ol, 1,2-benzisoxazol-3 2h-one, 1,2-benzoxazol-3-ol, 1,2-benzisoxazol-3-ol, 3-hydroxybenzisoxazole, 3-hydroxy-1,2-benzisoxazole, 1,2-benzisoxazolin-3-one, 1,2-benzisoxazol-3-ol 8ci, 2h-1,2-benzoxazol-3-one, benzisoxazolone
210830
1,2-benzoxazol-3-one
C1=CC=C2C(=C1)C(=O)NO2
CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 135.12
Percent Purity 95%
Chemical Name or Material Benzo[d]isoxazol-3-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'