missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carbohydrazide, 97%, Thermo Scientific™

Catalog Number p-2703680
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
175135-07-6
C9H8N2OS
192.24
MFCD00052501
ZXKPFIRPUUAAPQ-UHFFFAOYSA-N
benzo b thiophene-2-carbohydrazide, benzo b thiophene-2-carboxylic hydrazide, thianaphthene-2-carboxylic hydrazide, benzo b thiophene-2-carboxylicacid, hydrazide, maybridge1_008989, acmc-1c3go, #, benzo b thiophene-2-carboxylic acid hydrazide
519437
1-benzothiophene-2-carbohydrazide
NNC(=O)C1=CC2=CC=CC=C2S1
This item is not returnable. View return policy
175135-07-6
C9H8N2OS
192.24
MFCD00052501
ZXKPFIRPUUAAPQ-UHFFFAOYSA-N
benzo b thiophene-2-carbohydrazide, benzo b thiophene-2-carboxylic hydrazide, thianaphthene-2-carboxylic hydrazide, benzo b thiophene-2-carboxylicacid, hydrazide, maybridge1_008989, acmc-1c3go, #, benzo b thiophene-2-carboxylic acid hydrazide
519437
1-benzothiophene-2-carbohydrazide
NNC(=O)C1=CC2=CC=CC=C2S1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 192.24
Percent Purity 97%
Chemical Name or Material Benzo[b]thiophene-2-carbohydrazide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'