missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-carboxaldehyde, 97%, Maybridge™

Catalog Number p-3880563
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
4265-16-1
C9H6O2
146.145
ADDZHRRCUWNSCS-UHFFFAOYSA-N
benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural
61341
1-benzofuran-2-carbaldehyde
C1=CC=C2C(=C1)C=C(O2)C=O
This item is not returnable. View return policy
4265-16-1
C9H6O2
146.145
ADDZHRRCUWNSCS-UHFFFAOYSA-N
benzo b furan-2-carboxaldehyde, 2-benzofurancarboxaldehyde, benzofuran-2-carbaldehyde, benzofuran-2-carboxaldehyde, 2-formylbenzofuran, 2-benzofurancarbaldehyde, coumarilaldehyde, 1-benzofuran-2-carboxaldehyde, benzofuran-2-aldehyde, benzo b-2-furfural
61341
1-benzofuran-2-carbaldehyde
C1=CC=C2C(=C1)C=C(O2)C=O
CAS Min % 96.5
CAS Max % 100.0
Density 1.2060g/mL
Assay Percent Range 97%
Quantity 1 g
Formula Weight 146.14
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-2-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'