missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[a]pyrene, 96%, Thermo Scientific Chemicals
38.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mg
0.5 g
Description
Extensively used in cancer research and for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. It is also a multipurpose intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 177°C to 180°C |
Boiling Point | 495°C |
Quantity | 100 mg |
UN Number | UN3077 |
Merck Index | 14,1103 |
Solubility Information | Soluble in benzene, toluene, and xylene; Sparingly soluble in alcohol, methanol; Insoluble in water |
Formula Weight | 252.3 |
Percent Purity | 96% |
Physical Form | Crystalline |
Chemical Name or Material | Benzo[a]pyrene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'