missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems BenzNuclease DNA and RNA Nuclease (Ultra Pure,Protease Free)
Click to view available options
Quantity:
10 kU
50 kU
500 kU
Unit Size:
10ku
500ku
50ku
Description
BenzNuclease is a recombinant form of Serratia macescens extracellular endonuclease produced in Escherichia coli cells using a proprietary process at ACRObiosystems. BenzNuclease is a homodimer with monomer molecular masses about 30 kDa. Two disulfide bon
Specifications
Specifications
| Purity | >95% as determined by SDS-PAGE |
| Storage Buffer | Tris HCl, pH 8.0, MgCl2, and NaCl |
| Content And Storage | -20°C |
| Expression System | E. coli |
| Gene Alias | DNA and RNA Nuclease,BenzNuclease |
| Protein Tag | Tag Free |
| Shipping Condition | RT |
| Conjugate | Unconjugated |
| Source | BenzNuclease-DNA-RNA-Nuclease (BEE-N3116) is expressed from E.coli cells. |
| Quantity | 50 kU |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction