Learn More
Benzil monohydrazone, 98%, ACROS Organics™
Brand: Acros Organics 105430050
Code : 29
Additional Details : CAS Number : 5344-88-7 Weight : 0.06000kg
Quantity | 5g |
---|---|
Packaging | Glass Bottle |
Chemical Identifiers
Benzil monohydrazone | |
5344-88-7 | |
100.0 | |
98% | |
C6H5C(=NNH2)COC6H5 | |
MFCD00007625 |
98% | |
97.5 | |
98% | |
C14H12N2O | |
224.26 |
Specifications
Benzil monohydrazone | |
5344-88-7 | |
100.0 | |
Glass Bottle | |
98% | |
C14H12N2O | |
224.26 | |
147.0° to 151.0°C | |
5g |
98% | |
97.5 | |
Authentic | |
Very Fine Crystalline Powder | |
98% | |
C6H5C(=NNH2)COC6H5 | |
MFCD00007625 | |
Very slightly yellow |