missing translation for 'onlineSavingsMsg'
Learn More

Benzhydrylidenefluorene, 98+%, Thermo Scientific™

Catalog Number p-7027732
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
4709-68-6
C26H18
330.43
MFCD00051452
CGUOPWAERAISDV-UHFFFAOYSA-N
fluorene, 9-diphenylmethylene, 9-diphenylmethylene-9h-fluorene, 9-diphenylmethylene fluorene, 1,1-diphenyl-2-biphenyleneethylene, 9h-fluorene, 9-diphenylmethylene, 9-diphenylmethylidene fluorene, benzhydrylidene fluorene, 9-diphenylmethylene-9h-fluorene #
9-(diphenylmethylidene)-9H-fluorene
C1=CC=C(C=C1)C(=C1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1
This item is not returnable. View return policy
4709-68-6
C26H18
330.43
MFCD00051452
CGUOPWAERAISDV-UHFFFAOYSA-N
fluorene, 9-diphenylmethylene, 9-diphenylmethylene-9h-fluorene, 9-diphenylmethylene fluorene, 1,1-diphenyl-2-biphenyleneethylene, 9h-fluorene, 9-diphenylmethylene, 9-diphenylmethylidene fluorene, benzhydrylidene fluorene, 9-diphenylmethylene-9h-fluorene #
9-(diphenylmethylidene)-9H-fluorene
C1=CC=C(C=C1)C(=C1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 230°C to 232°C
Quantity 1 g
Beilstein 1977991
Formula Weight 330.43
Percent Purity ≥98%
Chemical Name or Material Benzhydrylidenefluorene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'