missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Benzhydryl alcohol, polymer supported, 1% cross-linked, 100-200 mesh, 0.3-2.0 mmol/g on poly(styrene-divinylbenzene)

Anchoring of carboxylic acids and alcohols

Brand:  Alfa Aesar™ 044454.14

Code : NEW

Additional Details : Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11420770

  • £578.23 / 25g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

Benzhydryl alcohol
Polymer supported, 1% cross-linked, 0.3 to 2.0 mmol/g on poly(styrene-divinylbenzene)
MFCD03702609

Specifications

Benzhydryl alcohol
Beads
Off-white to pink
100 to 200 Mesh
Polymer supported, 1% cross-linked, 0.3 to 2.0 mmol/g on poly(styrene-divinylbenzene)
MFCD03702609
25g
Videos
SDS