missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl bromide, 95%, Thermo Scientific™

Catalog Number p-4258755
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2297-65-6
C6H5BrO2S
221.07
CGWWQPZGKPHLBU-UHFFFAOYSA-N
phenylsulfonyl bromide
11138664
benzenesulfonyl bromide
C1=CC=C(C=C1)S(=O)(=O)Br
This item is not returnable. View return policy
2297-65-6
C6H5BrO2S
221.07
CGWWQPZGKPHLBU-UHFFFAOYSA-N
phenylsulfonyl bromide
11138664
benzenesulfonyl bromide
C1=CC=C(C=C1)S(=O)(=O)Br
CAS Min % 94.0
CAS Max % 100.0
Melting Point 19.5°C
Color Yellow to Brown
Boiling Point 83°C to 84°C (0.5 Torr)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 221.07
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Benzenesulfonyl bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzenesulfonyl bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'