missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic Acid Sodium Salt 98%, ACROS Organics™

Brand:  Acros Organics 401850025

UNSPSC : 12161500

Code : A0

Additional Details : CAS Number : 515-42-4 Weight : 2.50000kg

Quantity 2.5kg
Packaging Plastic drum

 View more versions of this product

Product Code. 13217059

  • £272.40 / 2.50kg


EU Stock 1
Add to basket

Specifications Description & Specifications

Chemical Identifiers

Benzenesulfonic acid, sodium salt
515-42-4
100.0
98%
SO3NaC6H5
MFCD00065179
98%
97.5
≥97.5%
C6H5NaO3S
180.16

Specifications

Benzenesulfonic acid, sodium salt
515-42-4
100.0
Authentic
Crystalline Powder
98%
SO3NaC6H5
MFCD00065179
2.5kg
98%
97.5
2% max. (105°C)
Plastic drum
≥97.5%
C6H5NaO3S
180.16
White to off-white

Safety and Handling

Product Identifier
  • Benzenesulfonic acid, sodium salt

Documentation

Technical Documents