missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzenesulfonic acid isopropyl ester, 95%, Thermo Scientific Chemicals
72.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
A sulfonate ester that acts as a potential effects in drug substances impurity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A sulfonate ester that acts as a potential effects in drug substances impurity.
Solubility
Soluble in chloroform and ethyl acetate.
Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
A sulfonate ester that acts as a potential effects in drug substances impurity.
Solubility
Soluble in chloroform and ethyl acetate.
Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Color | Colorless to Yellow |
Refractive Index | 1.502 |
Quantity | 5 g |
Beilstein | 1106314 |
Synonym | isopropyl benzenesulfonate, isopropylbenzenesulfonate, benzenesulfonic acid, 1-methylethyl ester, benzenesulfonic acid isopropyl ester, isopropyl besylate, benzenesulfonic acid,1-methylethyl ester, benzenesulfonic acid 1-methylethyl ester |
Solubility Information | Soluble in chloroform and ethyl acetate. |
Formula Weight | 200.25 |
Percent Purity | 95% |
Physical Form | Liquid |
Chemical Name or Material | Benzenesulfonic acid isopropyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'