Alfa Aesar™ Benzene-d6, 100% (Isotopic), contains 0.03% v/v TMS


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 042267.KV

BENZENE-D6 100% (ISOTOPIC)W 0.03% V/V TMS 10EA

UNSPSC: 12191501

Code: NEW

Additional Details:
Additional Details: CAS Number: 1076-43-3 Weight: 0.01000kg Transport: UN number : 1114 Chem class : 1 Pack group : II


Quantity 10 each

Product Code. 11349947

Quantity Price
1 £ 155.71 / Pack of 10
Estimated Shipment date
from Supplier 06-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Benzene-d_6
CAS 1076-43-3
Name Note Isotopic
Formula Weight 84.16
MDL Number MFCD00003010
UN Number UN1114

Specification

Formula Weight 84.16
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H225-H340-H350-H372-H304-H315-H319
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
Packaging Prescored Ampules
Chemical Name or Material Benzene-d_6
TSCA TSCA
Sensitivity Moisture Sensitive
Density 0.950g/mL
Name Note Isotopic
Boiling Point 79.1°C
Melting Point 6.8°C
Physical Form Liquid
Assay 100%
Quantity 10 each
Concentration contains 0.03% v/v TMS
Color Colorless
Refractive Index 1.4986
MDL Number MFCD00003010
CAS 1076-43-3
UN Number UN1114
Health Hazard 1 Danger
Odor Aromatic
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: BENZENE
Safety and Handling

Danger

GHS H Statement
H225-H340-H350-H372-H304-H315-H319
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.