Alfa Aesar™ Benzene-d6, 100% (Isotopic), contains 0.03% v/v TMS


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 042267.SA

UNSPSC: 12191501

Code: NEW

Additional Details:
Additional Details: CAS Number: 1076-43-3 Weight: 0.01000kg Transport: UN number : 1114 Chem class : 1 Pack group : II


Quantity 2 each

Product Code. 11339947

Quantity Price
1 £ 34.13 / Pack of 2
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Benzene-d_6
CAS 1076-43-3
Name Note Isotopic
Formula Weight 84.16
MDL Number MFCD00003010
UN Number UN1114

Specification

Formula Weight 84.16
Packaging Prescored Ampules
Health Hazard 2 GHS H Statement
H225-H340-H350-H372-H304-H315-H319
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 2 each
Density 0.950g/mL
CAS 1076-43-3
UN Number UN1114
Melting Point 6.8°C
Boiling Point 79.1°C
Color Colorless
Chemical Name or Material Benzene-d_6
Sensitivity Moisture Sensitive
Physical Form Liquid
TSCA TSCA
Refractive Index 1.4986
Name Note Isotopic
Health Hazard 1 Danger
Concentration contains 0.03% v/v TMS
MDL Number MFCD00003010
Odor Aromatic
DOT Information Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: BENZENE
Assay 100%
Safety and Handling

Danger

GHS H Statement
H225-H340-H350-H372-H304-H315-H319
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.