missing translation for 'onlineSavingsMsg'
Learn More

Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number p-4509495
22.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1670-14-
C7H9ClN2
156.61
MFCD00066285
LZCZIHQBSCVGRD-UHFFFAOYSA-N
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
16219042
Cl.NC(=N)C1=CC=CC=C1
This item is not returnable. View return policy
1670-14-
C7H9ClN2
156.61
MFCD00066285
LZCZIHQBSCVGRD-UHFFFAOYSA-N
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
16219042
Cl.NC(=N)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 167.0°C to 171.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C(NH2)NHC6H5·HCl
Quantity 5 g
Beilstein 09,28
Fieser 06,27
Solubility Information Solubility in water: soluble
IUPAC Name benzenecarboximidamide;hydrate;hydrochloride
Formula Weight 156.61
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Benzamidine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Benzamidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'