missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Benzaldoxime, predominantly (E)-isomer, 98%

Brand:  Alfa Aesar™ A12053.22

Code : NEW

Additional Details : CAS Number : 932-90-1 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11469932

  • £85.90 / 100g


Estimated Shipment date
from Supplier 22-01-2018
Add to basket

Description

Description

Applications
Oximes are used as a peel-preventing additive in paints and lacquers. It acts as an antioxidant against oxidative drying materials which forms sticky skin with air oxygen. Oximes are used as chemical building block for the synthesis of agrochemicals and pharmaceuticals. In medicine application, Oxime structure is effective in cholinesterase reactivators to treat the poisoning by organophosphates. Oxime is used as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.

Solubility
Slightly soluble in water. Soluble in ethanol and ether.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases, acids, acid anhydrides and acid chlorides.
Specifications

Chemical Identifiers

Benzaldoxime, predominantly (E)-isomer
C7H7NO
MFCD00002119
Benzaldehyde oxime
932-90-1
121.14
774138

Specifications

Benzaldoxime, predominantly (E)-isomer
98%
121.14
774138
33° to 35°C
100g
932-90-1
C7H7NO
MFCD00002119
Benzaldehyde oxime
118° to 120°C (10mmHg)
108°C (226°F)
Videos
SDS