missing translation for 'onlineSavingsMsg'
Learn More

Bentonite, Acros Organics

Brand:  Acros Organics 447440025

Code : 29

Additional Details : CAS Number : 1302-78-9 Weight : 2.55000kg

Quantity 2.5kg
Packaging Plastic bottle

 View more versions of this product

Product Code. 11934061

  • £80.20 / 2.50kg


Estimated Shipment
08-06-2018
Add to basket

Description

Description

Drying agent, Lewis acid
Specifications

Chemical Identifiers

Bentonite
1302-78-9

Specifications

Bentonite
1302-78-9
Plastic bottle
2.5kg
Videos
SDS