Learn More
Lenz Laborglasinstrumente™ Bent Receiver Adapter with Glass Olive
Allows for the connection of a condenser to a receiving flask. Lenz Laborglasinstrumente™ Bent Receiver Adapters with Glass Olive can be directly heated and withstand typical laboratory thermal variations.
Brand: Lenz Laborglasinstrumente™ 05275001
Code : A0
Additional Details : Weight : 0.02700kg
Description
- NS socket and NS cone
- 105° angle
- Vacuum connection
- Glass olive with drain tip
- Meets DIN 12594 standards
Specifications
Bent Receiver Adapter with Glass Olive | |
Duran Glass | |
DIN 12594 | |
NS 14/23 |