missing translation for 'onlineSavingsMsg'
Learn More

Benidipine hydrochloride, Tocris Bioscience™

Calcium channel blocker

Brand:  Tocris (Bio-Techne) 393410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13443834

  • £139.12 / 10mg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Orally active antihypertensive agent which displays a wide range of activities in vitro and in vivo. Inhibits L-, N- and T-type Ca2+ channels. Also inhibits aldosterone-induced mineralocorticoid receptor activation. Exhibits cardioprotective and antiartherosclerotic effects.
Specifications

Chemical Identifiers

91599-74-5
542.029
(4R)-rel-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 3-methyl 5-[(3R)-1-(phenylmethyl)-3-piperidinyl] ester hydrochloride
5-O-[(3R)-1-benzylpiperidin-3-yl] 3-O-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
C28H32ClN3O6
KILKDKRQBYMKQX-MIPPOABVSA-N
656667
CC1=C(C(C(=C(N1)C)C(=O)OC2CCCN(C2)CC3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl

Specifications

Benidipine hydrochloride
91599-74-5
(4R)-rel-1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 3-methyl 5-[(3R)-1-(phenylmethyl)-3-piperidinyl] ester hydrochloride
CC1=C(C(C(=C(N1)C)C(=O)OC2CCCN(C2)CC3=CC=CC=C3)C4=CC(=CC=C4)[N+](=O)[O-])C(=O)OC.Cl
542.029
542.02
10mg
C28H32ClN3O6
KILKDKRQBYMKQX-MIPPOABVSA-N
5-O-[(3R)-1-benzylpiperidin-3-yl] 3-O-methyl (4R)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
656667
>99%
Videos
Documents