missing translation for 'onlineSavingsMsg'
Learn More

Bendamustine hydrochloride, Tocris Bioscience™

Cytostatic agent; exhibits DNA alkylating and purine analog properties

Brand:  Tocris (Bio-Techne) 368110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13453414

  • £133.77 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cytostatic agent that displays activity in non-Hodgkin's lymphomas. Exhibits bifunctionality; combines DNA alkylating properties with those of purine analogs.
Specifications

Chemical Identifiers

3543-75-7
394.721
bendamustine hydrochloride, treanda, bendamustine hcl, ribomustin, cytostasan, bendamustin hydrochloride, treakisym, 4-5-bis 2-chloroethyl amino-1-methyl-1h-benzo d imidazol-2-yl butanoic acid hydrochloride, bendeka
4-[5-[bis(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoic acid;hydrochloride
C16H22Cl3N3O2
ZHSKUOZOLHMKEA-UHFFFAOYSA-N
77082
CN1C2=C(C=C(C=C2)N(CCCl)CCCl)N=C1CCCC(=O)O.Cl

Specifications

10mg
3543-75-7
bendamustine hydrochloride, treanda, bendamustine hcl, ribomustin, cytostasan, bendamustin hydrochloride, treakisym, 4-5-bis 2-chloroethyl amino-1-methyl-1h-benzo d imidazol-2-yl butanoic acid hydrochloride, bendeka
CN1C2=C(C=C(C=C2)N(CCCl)CCCl)N=C1CCCC(=O)O.Cl
394.721
394.72
Bendamustine hydrochloride
C16H22Cl3N3O2
ZHSKUOZOLHMKEA-UHFFFAOYSA-N
4-[5-[bis(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoic acid;hydrochloride
77082
>98%
Videos
Documents