missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Benchtop Microcentrifuge

Choose from a range of models with different rotors for a variety of centrifugation applications. Eppendorf™ Benchtop Microcentrifuges are ideal for safe centrifugation of hazardous samples.

Brand:  Eppendorf™ 5404000464

Code : 99

Additional Details : Weight : 25.00000kg

 View more versions of this product

Product Code. 10755822

  • £4451.00 / Each

In Stock 5

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Standard rotor can accommodate 1.5mL/2.0mL tubes
  • PCR* strips and kits/spin columns can be centrifuged in the corresponding rotor for molecular and biological laboratories
  • Automatic lid opening at the end of the run – prevents sample warming and allows easy access to samples (Centrifuge 5418 and 5424)
The 5418 and 5418 R models are 18 place microcentrifuges for 1.5mL to 2.0mL tubes.
Small footprint
  • Quiet operation without rotor lid
  • Aerosol-tight rotor as standard
  • Converts rpm/RCF
  • Short spin key For research and clinical laboratories
Centrifuge 5424 and 5424 R feature 24–place capacity and speed up to 21,130 × g
  • Available as a rotary knob version for fast parameter selection, or keypad version for easy cleaning
  • OptiBowl design for quiet operation, even without a rotor lid
  • Soft-touch one-finger lid closure for ergonomic operation
Specifications

Specifications

21kg
21,130 x G
<54 dB(A), with rotor lid: <48 dB(A) with Rotor FA-45-24-11
Benchtop Microcentrifuge
Aerosol-tight
Knob
350w
Yes
15sec.
30 s to 9:59 hr.
24 x 1.5/2mL
Digital
–10 to 40°C
Rotor
230V 50/60Hz
480mm
100 to 15,000rpm
Yes
Fixed angle
5424R
Videos
Documents
Special Offers

Special Offers

  • Eppendorf , Eppendorf New Brunswick
    Simply better pipetting
    : 13837

    2019-12-31

    Discount will apply to qualifying orders received by Fisher Scientific between 01/09/2019 and 31/12/2019. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions