missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ X-Ray Cassette Security Bags
Intended to augment the light protection afforded by X-ray film cassettes which may leak light because of damage due to age, use, or sudden shock
POA
Specifications
For Use With (Equipment) | X-ray Film Cassette |
---|---|
Product Type | Security Bag |
Material | Nylon |
Color | Black |
For Use With (Application) | Used for additional protection from light for X-ray film cassettes that you are currently using |
Description
- Made of black nylon
- Hook-and-loop sealing strip on its closing flap
Specifications
X-ray Film Cassette | |
Nylon | |
Used for additional protection from light for X-ray film cassettes that you are currently using |
Security Bag | |
Black |