missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Scienceware™ Switch-Grid™ Test Tube Racks

One rack provides 25 possible tube size combinations

Brand:  Bel-Art™ F18745-0014

Code : 04

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 13482459

  • £21.75 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Innovative, split-frame rack holds up to two different size tubes in the same rack, allowing users to quickly switch grids to hold other sizes as needed.

  • 10mm to 30mm diameter test tubes fit in 5 colored coded grid inserts
  • Three-tier rack holds tubes securely in place
  • Rubber feet keep racks from sliding on benchtop; provide secure stacking while in storage
  • Chemical resistant polyoxymethylene plastic rack frame and grids can be cleaned with mild detergent, ethanol or isopropyl alcohol; autoclaved at 121°C (250°F)
  • More compact and less expensive than purchasing multiple racks
  • Rack frame overall dimensions: 31.8L x 11.1W x 6.7cmH (12 1/2 x 4 3/8 x 2 5/8 in.)

Specifications

Specifications

Yellow
6.7cm
31.8cm
12
11.1cm
12.5 x 4.37 x 2.62 in. (31.8 x 11.1 x 6.7cm)
25 to 30mm Tubes
Plastic
Rows: 6x2
Rack with Yellow Grids
Videos