Learn More
Bel-Art™ SP Scienceware™ Super Biohazard Disposal Bags
Protects against dangerous tears and leaks. SP Scienceware™ Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.
Brand: Bel-Art™ 06494-00
Code : A0
Additional Details : Weight : 2.35800kg
Description
- Protect against dangerous tears and leaks
- Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
- Extra-heavy-duty bags are 2mil-thick blended polymer
- Autoclavable to 121°C (250°F)
- Sterilization indicator patch darkens when exposed to steam sterilization