missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Super Biohazard Disposal Bags

Protects against dangerous tears and leaks. SP Scienceware™ Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.

Brand:  Bel-Art™ 06494-02

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 10683503

  • £400.74 / Pack of 200


Estimated Shipment
21-05-2018
Add to basket

Description

Description

 

  • Protect against dangerous tears and leaks
  • Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
  • Extra-heavy-duty bags are 2mil-thick blended polymer
  • Autoclavable to 121°C (250°F)
  • Sterilization indicator patch darkens when exposed to steam sterilization

 

Specifications

Specifications

23 x 19 in. (58 x 48cm)
Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
0.279mm
Orange Red
Polymer
Extra-strength, high-impact, high-temperature bags reduce autoclave time
Biohazard Disposal Bag
Yes
58cm
48cm
Videos