missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Stir and Add™ Cuvet Mixer
Made of clear acrylic with a straight handle, this tool is used when reactions are performed directly into cuvettes, to mix added reagents
Brand: Bel-Art™ F37752-0000
Code : 03
Additional Details : Weight : 0.01000kg
Description
- Up and down motion in cell provides complete mixing
- Shaft diameter 3.17mm (0.125 in.)
- Straight handle
Used to add enzymes and other materials to solution in spectrophotometer cells.
Specifications
3.17 mm | |
0.125 in. | |
Used to add enzymes and other materials to solution in spectrophotometer cells, this tool is used when reactions are performed directly into cuvettes, to mix added reagents | |
Acrylic |
4.5 in. | |
Cell type: 10mm light path; Straight handle | |
36/Cs. | |
11.4 cm |