Learn More
Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainer
Resistant to chemicals sink strainer has four strong magnets in each corner to attract and hold Spinbar magnetic stirring bars. Bel Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainer is a simple solution to prevent your Spinbar™ Magnetic Stirring Bars from going down the sink drain.
Brand: Bel-Art™ F37787-0001
Code : 03
Additional Details : Weight : 0.11092kg
Description
- Long slits in the sides and skirt allow fluids to drain easily and rapidly
- A wide skirt enables strainer to be used in a variety of sink drains; flat edge on one side allows it to be placed close to the sink wall
- Four legs on the underside anchor the strainer firmly in sink drain and prevent it from being dislodged by flowing liquid
- Color choices improve visibility of stirring bars
Specifications
4 strong magnets in each corner, long slits in the sides and wide skirts | |
Spinbar™ Magnetic Sink Strainer | |
Stirring bars |
Prevents stir bar from washing down drain | |
Green |