missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models

Constructed of Durastar™ co-polyester. Bel Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Vertical Models block 99% of UV light and are resistant to staining, crazing and chemical attack.

Brand:  Bel-Art™ F42074-1008

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11836890

  • £614.17 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Amber color models reduce visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
  • Compact 34.1W x 41.4cmD footprint for space saving stacking up to 3 units high
  • Replacement shelves available; 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves
  • Can be used with desiccant cartridges, F420450250, F420450200, F420480100
Specifications

Specifications

Amber
16.3 x 13.4 in. (41.4 x 34.1cm)
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250
41.4cm
Chemicals, stains and fine cracks
34.1cm
Copolyester
51.8cm
41.4cm
Left-hinged and gasketed
Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
4.0
Removable perforated shelves
Vertical
No
3
Videos