missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Secador™ Auto Desiccator Cabinets: Vertical Models

A UV light blocking, dry environment with automatic electronic dehumidification

Brand:  Bel-Art™ F42074-1220

Code : Z1

Additional Details : Weight : 9.50000kg

 View more versions of this product

Product Code. 11802742

  • £1345.00 / Each
Estimated Shipment: 30-05-2024
to see stock.
Add to basket

Description

Description

A UV light blocking, dry environment with automatic electronic dehumidification

The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.

  • Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants
  • Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
  • Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
  • U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626

  • 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info)
  • Dimensions
    • Secador 2.0: 1.16 cu. ft. internal volume, 16.3L × 13.4W × 12.4 in. H
    • Secador 3.0: 1.55 cu. ft. internal volume, 16.3L × 13.4W × 16.4 in. H
    • Secador 4.0: 1.87 cu. ft. internal volume, 16.3L × 13.4W × 20.4 in. H

    Order Info

    Replacement shelves are available.

    Specifications

    Specifications

    Not Autoclavable
    Vertical
    Side-swinging
    Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
    3
    Vented Adjustable
    5°C to 40°C
    16.3 x 13.4 in. (41.4 x 34.1 cm)
    41.4 cm
    8.9 in.
    22.5 cm
    16.4 in.
    16.3 in.
    230 V
    Clear
    Copolyester
    Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant
    4.0
    Chemicals, stains and fine cracks
    41°F to 104°F
    1.87 cu. ft.
    16.3 in.
    13.4 in.
    34.1 cm
    20.4 in.
    51.8 cm
    41.4 cm
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    View accessory products
    Special Offers

    Special Offers