Learn More
Bel-Art™ Secador™ Auto Desiccator Cabinets: Vertical Models
A UV light blocking, dry environment with automatic electronic dehumidification
Brand: Bel-Art™ F42072-1228
Code : 03
Additional Details : Weight : 8.40000kg
Description
A UV light blocking, dry environment with automatic electronic dehumidification
The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
- Patented electronic dehumidification system reduces humidity and circulates dry air automatically, freeing users from monitoring and changing desiccants
- Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
- Amber color models reduce visible light penetration by over 50% for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors with easy-to-view dial hygrometer maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
- U.S. Patents: 6,772,534; 6,834,920; 7,114,266; 7,318,630 and D474,626
- 4.0 units can be converted from vertical to horizontal configuration or vice versa by turning the unit and ordering the appropriate number of shelves (see Ordering Info)
- Secador 2.0: 1.16 cu. ft. internal volume, 16.3L × 13.4W × 12.4 in. H
- Secador 3.0: 1.55 cu. ft. internal volume, 16.3L × 13.4W × 16.4 in. H
- Secador 4.0: 1.87 cu. ft. internal volume, 16.3L × 13.4W × 20.4 in. H
Order Info
Replacement shelves are available.
Specifications
Not Autoclavable | |
Vertical, Tinted | |
Side-swinging | |
Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer | |
2 | |
Perforated | |
5°C to 40°C | |
16.3 x 13.4 in. (41.4 x 34.1 cm) | |
41.4 cm | |
34.1 cm | |
31.4 cm | |
41.4 cm |
Amber | |
Copolyester | |
Easy and reliable storage of moisture and light sensitive products, can be used with Scienceware™ desiccant cartridges, canisters or similar desiccant | |
2.0 | |
Chemicals, stains and fine cracks | |
41°F to 104°F | |
1.16 cu. ft. | |
16.3 in. | |
13.4 in. | |
12.4 in. | |
16.3 in. | |
230 V |