Learn More
Bel-Art™ Poxygrid™ Wire Test Tube Racks
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
Brand: Bel-Art™ F18757-0000
Code : Z1
Additional Details : Weight : 1.00000kg
Description
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores from 40 to 108 tubes (five different diameters of testtubes)
- Autoclavable
Specifications
Test Tube Rack | |
Green | |
Epoxy Coated Steel | |
9.5 x 5.1 x 2.5 in. (242 x 130 x 64mm) | |
9.48 in. | |
5.12 in. | |
2.51 in. |
Autoclavable | |
Tubes (13 to 16 mm) | |
72 | |
242 mm | |
130 mm | |
64 mm | |
16 mm |