Learn More
Bel-Art™ SP Scienceware™ Polypropylene Sterilizing Trays
Corrosion-proof polypropylene plastic trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.
Brand: Bel-Art™ H16264-0000
Code : A0
Additional Details : Weight : 0.78000kg
Description
Corrosion-proof polypropylene plastic trays. Bel Art™ SP Scienceware™ Polypropylene Sterilizing Trays have rounded corners for easy cleaning and smooth, tapered sides for stacking.
- Ideal for sterilizing instruments, bottles containing agar, deionized water, saline and other reagents, and for safely cleaning glassware in sinks
- Trays are steam autoclavable at 121°C (250°F)
- Covers are sold separately
Specifications
18 L | |
12.7 cm | |
Polypropylene | |
39.4 cm | |
21 in. | |
Rectangular | |
Autoclavable |
20.3 x 15.5 x 5 in. (53.3 x 39.4 x 12.7 cm) | |
53.3 cm | |
Sterilizing Tray | |
Fits 18L Tray Cover, Mfr. No. H162590000 | |
5 in. | |
15.5 in. |