missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ PLUM™ Emergency Eyewash Station Refills

Refill kits for PLUM station

Brand:  Bel-Art™ F24880-1023

Code : Z1

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 12729706

  • £34.55 / Pack of 3


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Bottles contain a sterile 0.9% sodium chloride solution (saline) for rinsing dust and debris, or a 4.9% sterile phosphate buffer (pH neutralizing) solution to quickly neutralize acids and alkalis
  • Bottles have clear first aid instructions, twist-open design for quick access and a built-in eye cup with dust cap
  • Use to refill eye wash stations or take to job sites as part of first aid kits
  • Bottles require no maintenance and have a three-year shelf life from date of manufacture

Specifications

Specifications

200mL
PLUM eyewash stations or alone
pH neutralizing eyewash solution refill
6.76 oz., 200mL
Three 200mL bottles of sterile pH neutralizaing solution form-fitting eye cup, a dust cap and clear instructions
Videos