missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Minipet™ Pipettors

The chemical resistance of PTFE resin in a self-filling, syringe-type repetitive dispenser

Brand:  Bel-Art™ F37900-0100

Code : NEW

Additional Details : CAS Number : 60669-69-4 Weight : 0.01000kg

 View more versions of this product

Product Code. 10043453

  • £342.25 / Each


Estimated Shipment
05-07-2021
Add to basket

Description

Description

  • Features a proprietary PTFE 3-way valve and fittings on a borosilicate glass barrel
  • Self-locking PTFE plunger adjusts over full volume range of the syringe, yet won't move from original setting even after multiple dispensing
  • Safe for use with virtually any solvent or acid except HF
  • Autoclavable and suited for dispensing media and other sterile samples
  • Once the 18 in. long (20.3cm) PTFE filling tube is placed in a liquid reservoir, reagent flows through a closed system
  • Accuracy: ±0.5%
  • Molded plastic handle is ergonomically designed to minimize operator fatigue when used for extended periods

Order Info

Complete with filling tube and sinker.

Specifications

Specifications

±0.5%
PTFE 3-way check valve Teflon-coated plunger, ergonomic Polypropylene handle
1
Autoclavable
0.2mL
10 mL
Videos
SDS
Documents