missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Minipet™ Pipettors
The chemical resistance of PTFE resin in a self-filling, syringe-type repetitive dispenser
£349.10
Specifications
Accuracy | ±0.5% |
---|---|
Autoclavable | Autoclavable |
For Use With | Dispensing Media and Other Sterile Samples |
Number of Channels | 1 |
Type | Pipette |
Product Code | Brand | Volume (Metric) | Increments | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Increments | Price | Quantity & Availability | ||||
10043453
|
Bel-Art™
F37900-0100 |
10 mL | 0.2mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Features a proprietary PTFE 3-way valve and fittings on a borosilicate glass barrel
- Self-locking PTFE plunger adjusts over full volume range of the syringe, yet won't move from original setting even after multiple dispensing
- Safe for use with virtually any solvent or acid except HF
- Autoclavable and suited for dispensing media and other sterile samples
- Once the 18 in. long (20.3cm) PTFE filling tube is placed in a liquid reservoir, reagent flows through a closed system
- Accuracy: ±0.5%
- Molded plastic handle is ergonomically designed to minimize operator fatigue when used for extended periods
Order Info
Complete with filling tube and sinker.
Specifications
±0.5% | |
Dispensing Media and Other Sterile Samples | |
Pipette |
Autoclavable | |
1 | |
PTFE 3-way check valve Teflon-coated plunger, ergonomic Polypropylene handle |